ENA048303275 Enamine

2-hydroxy-3-methoxybenzamide

  • MDL number MFCD06357904
  • Cas Number : 26751-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11066
Availability details for
Quantity
  • Availability
  • Add to Cart