ENA048303279 Enamine

3-(5,6,7,8-tetrahydronaphthalene-2-sulfonamido)propanoic acid

  • MDL number MFCD06358663
  • Cas Number : 790681-74-2
  • Molecular Weight : 283.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11073
Availability details for
Quantity
  • Availability
  • Add to Cart