ENA048303281 Enamine

2-chloro-1-[1-(4-fluorobenzyl)-2,5-dimethyl-1H-pyrrol-3-yl]ethanone

  • MDL number MFCD06358661
  • Cas Number : 790681-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11077

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11077
Availability details for
Quantity
  • Availability
  • Add to Cart