ENA048303283 Enamine

[(5,6,7,8-tetrahydronaphthalen-2-ylsulfonyl)amino]acetic acid

  • MDL number MFCD06358665
  • Cas Number : 790681-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11081

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11081
Availability details for
Quantity
  • Availability
  • Add to Cart