ENA048303292 Enamine

2-chloro-N-[3-(4-ethylphenyl)-1-phenyl-1H-pyrazol-5-yl]acetamide

  • MDL number MFCD06358649
  • Cas Number : 956453-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11092
Availability details for
Quantity
  • Availability
  • Add to Cart