ENA048303297 Enamine

5-(morpholine-4-sulfonyl)-2-(pyrrolidin-1-yl)benzoic acid

  • MDL number MFCD06357881
  • Cas Number : 790271-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11101
Availability details for
Quantity
  • Availability
  • Add to Cart