ENA048303301 Enamine

N-[4-(isothiocyanatomethyl)-1,3-thiazol-2-yl]-N-mesitylacetamide

  • MDL number MFCD06358641
  • Cas Number : 790681-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11107

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11107
Availability details for
Quantity
  • Availability
  • Add to Cart