ENA048303302 Enamine

N-(2-chloro-4,6-dimethylphenyl)-N-[4-(isothiocyanatomethyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD06358642
  • Cas Number : 790681-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11108

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11108
Availability details for
Quantity
  • Availability
  • Add to Cart