ENA048303312 Enamine

3-(4-cyclohexylphenyl)-1-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD06358655
  • Cas Number : 956159-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11126

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11126
Availability details for
Quantity
  • Availability
  • Add to Cart