ENA048303321 Enamine

N-[3-(4-butylphenyl)-1-phenyl-1H-pyrazol-5-yl]-2-chloroacetamide

  • MDL number MFCD06359956
  • Cas Number : 956411-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11160
Availability details for
Quantity
  • Availability
  • Add to Cart