ENA048303322 Enamine

({[(4-fluorophenyl)sulfonyl]amino}oxy)acetic acid

  • MDL number MFCD06359955
  • Cas Number : 790725-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11161

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11161
Availability details for
Quantity
  • Availability
  • Add to Cart