ENA048303326 Enamine

3-({5-[(4-methoxyphenyl)sulfamoyl]-2-(morpholin-4-yl)phenyl}carbamoyl)prop-2-enoic acid

  • Cas Number : 790725-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11166
Availability details for
Quantity
  • Availability
  • Add to Cart