ENA048303327 Enamine

1-{[4-(trifluoromethyl)phenyl]sulfonyl}piperazine

  • MDL number MFCD06363488
  • Cas Number : 793679-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11167

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-11167
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart