ENA048303336 Enamine

2-chloro-N-[3-(2,3-dihydro-1H-inden-5-yl)-1-phenyl-1H-pyrazol-5-yl]acetamide

  • MDL number MFCD06360000
  • Cas Number : 956262-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11180
Availability details for
Quantity
  • Availability
  • Add to Cart