ENA048303338 Enamine

4-(chloromethyl)-2-(thiophen-2-yl)-1,3-oxazole

  • MDL number MFCD06359997
  • Cas Number : 54679-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-57
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11182
Availability details for
Quantity
  • Availability
  • Add to Cart