ENA048303344 Enamine

2-chloro-N-[5-chloro-2-(1H-1,2,4-triazol-1-yl)phenyl]acetamide

  • MDL number MFCD06362450
  • Cas Number : 792953-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11197
Availability details for
Quantity
  • Availability
  • Add to Cart