ENA048303346 Enamine

10-(chloroacetyl)-2-(methylthio)-10H-phenothiazine

  • MDL number MFCD06362447
  • Cas Number : 65010-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11201

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11201
Availability details for
Quantity
  • Availability
  • Add to Cart