ENA048303348 Enamine

6-amino-1-isobutyl-5-[(3-methoxypropyl)amino]pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06361945
  • Cas Number : 791600-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11206

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11206
Availability details for
Quantity
  • Availability
  • Add to Cart