ENA048303349 Enamine

2,2-dioxo-3H,4H-2lambda6-pyrazino[2,1-c][1,2,4]thiadiazine-9-carboxylic acid

  • MDL number MFCD06364463
  • Cas Number : 793716-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11209
Availability details for
Quantity
  • Availability
  • Add to Cart