ENA048303352 Enamine

2-chloro-N-[3-(4-ethoxybenzoyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophen-2-yl]acetamide

  • MDL number MFCD06362475
  • Cas Number : 792954-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11222
Availability details for
Quantity
  • Availability
  • Add to Cart