ENA048303354 Enamine

N-[3-(4-tert-butylbenzoyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophen-2-yl]-2-chloroacetamide

  • MDL number MFCD06362473
  • Cas Number : 792954-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11224
Availability details for
Quantity
  • Availability
  • Add to Cart