ENA048303372 Enamine

[(4-chlorophenyl)methyl][1-(thiophen-2-yl)propan-2-yl]amine

  • MDL number MFCD06361977
  • Cas Number : 791601-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11256

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11256
Availability details for
Quantity
  • Availability
  • Add to Cart