ENA048303375 Enamine

benzyl[1-(thiophen-2-yl)propan-2-yl]amine

  • MDL number MFCD06361976
  • Cas Number : 791601-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11266
Availability details for
Quantity
  • Availability
  • Add to Cart