ENA048303378 Enamine

(2-amino-4,5,6,7-tetrahydro-1-benzothien-3-yl)(3,5-dimethoxyphenyl)methanone

  • MDL number MFCD06363449
  • Cas Number : 793678-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11272
Availability details for
Quantity
  • Availability
  • Add to Cart