ENA048303379 Enamine

3-(3-fluorobenzoyl)-4,5,6,7-tetrahydro-1-benzothiophen-2-amine

  • MDL number MFCD06363450
  • Cas Number : 399043-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11274
Availability details for
Quantity
  • Availability
  • Add to Cart