ENA048303383 Enamine

(2-amino-6-tert-butyl-4,5,6,7-tetrahydro-1-benzothien-3-yl)(3-fluorophenyl)methanone

  • MDL number MFCD06363454
  • Cas Number : 793678-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11278

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11278
Availability details for
Quantity
  • Availability
  • Add to Cart