ENA048303384 Enamine

(2-Amino-6-tert-butyl-4,5,6,7-tetrahydro-benzo[b]thiophen-3-yl)-(3,5-dimethoxy-phenyl)-methanone

  • MDL number MFCD06363455
  • Cas Number : 793678-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11279

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11279
Availability details for
Quantity
  • Availability
  • Add to Cart