ENA048303397 Enamine

1-(2,4-dimethylphenyl)-5-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD06363467
  • Cas Number : 793678-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11294
Availability details for
Quantity
  • Availability
  • Add to Cart