ENA048303411 Enamine

6-({[4-(pyrrolidin-1-ylsulfonyl)phenyl]sulfonyl}amino)hexanoic acid

  • MDL number MFCD06363741
  • Cas Number : 793690-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11319
Availability details for
Quantity
  • Availability
  • Add to Cart