ENA048303412 Enamine

5-[(diphenylmethyl)sulfamoyl]-2-methylbenzoic acid

  • MDL number MFCD06364489
  • Cas Number : 793716-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11325
Availability details for
Quantity
  • Availability
  • Add to Cart