ENA048303413 Enamine

N-(4-aminophenyl)-2,6-dichlorobenzene-1-sulfonamide

  • MDL number MFCD06364460
  • Cas Number : 793715-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11327
Availability details for
Quantity
  • Availability
  • Add to Cart