ENA048303415 Enamine

4-ethoxy-3-(pyrrolidin-1-ylsulfonyl)aniline

  • MDL number MFCD06363474
  • Cas Number : 793678-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11329
Availability details for
Quantity
  • Availability
  • Add to Cart