ENA048303416 Enamine

3-(1-propyl-5-sulfamoyl-1H-1,3-benzodiazol-2-yl)propanoic acid

  • MDL number MFCD06363475
  • Cas Number : 793678-98-5
  • Molecular Weight : 311.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-231
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11330
Availability details for
Quantity
  • Availability
  • Add to Cart