ENA048303418 Enamine

3-(3,5-dimethoxybenzoyl)-6-methyl-4,5,6,7-tetrahydro-1-benzothiophen-2-amine

  • MDL number MFCD06364482
  • Cas Number : 793716-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11333
Availability details for
Quantity
  • Availability
  • Add to Cart