ENA048303423 Enamine

5-(4-bromophenyl)-1-(3-isopropoxypropyl)-1H-imidazole-2-thiol

  • MDL number MFCD06364476
  • Cas Number : 793716-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11340
Availability details for
Quantity
  • Availability
  • Add to Cart