ENA048303429 Enamine

4-imino-4H-3,1-benzothiazine-2-thiol

  • MDL number MFCD06364471
  • Cas Number : 793716-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11349

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11349
Availability details for
Quantity
  • Availability
  • Add to Cart