ENA048303430 Enamine

5-ethyl-3-(4-methoxyphenyl)-6-methyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06364468
  • Cas Number : 793716-04-8
  • Molecular Weight : 332.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11350
Availability details for
Quantity
  • Availability
  • Add to Cart