ENA048303433 Enamine

5-(piperidine-1-sulfonyl)-2-(pyrrolidin-1-yl)benzoic acid

  • MDL number MFCD06364943
  • Cas Number : 793727-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11359
Availability details for
Quantity
  • Availability
  • Add to Cart