ENA048303444 Enamine

1-(5-chloro-2,4-dimethoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD06364961
  • Cas Number : 793727-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11376
Availability details for
Quantity
  • Availability
  • Add to Cart