ENA048303451 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]-2,2-dimethylpropanamide

  • MDL number MFCD06368761
  • Cas Number : 795290-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11389

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11389
Availability details for
Quantity
  • Availability
  • Add to Cart