ENA048303459 Enamine

2-chloro-N-{4-[(2,5-dimethylphenyl)sulfanyl]phenyl}acetamide

  • MDL number MFCD06365001
  • Cas Number : 793727-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11401
Availability details for
Quantity
  • Availability
  • Add to Cart