ENA048303467 Enamine

1-(4-methylphenyl)-5-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD06364989
  • Cas Number : 136802-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11419
Availability details for
Quantity
  • Availability
  • Add to Cart