ENA048303468 Enamine

1-benzyl-3-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06364990
  • Cas Number : 905589-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-158
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11420
Availability details for
Quantity
  • Availability
  • Add to Cart