ENA048303475 Enamine

1-(2,4-dimethylphenyl)-5-(4-methoxyphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD06366738
  • Cas Number : 794554-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11440

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11440
Availability details for
Quantity
  • Availability
  • Add to Cart