ENA048303477 Enamine

(2E)-3-(1-benzyl-3-phenyl-1H-pyrazol-4-yl)acrylic acid

  • MDL number MFCD06364959
  • Cas Number : 1006494-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11445
Availability details for
Quantity
  • Availability
  • Add to Cart