ENA048303478 Enamine

3-(2-chloroacetyl)-1-(1,1-dioxo-1lambda6-thiolan-3-yl)urea

  • MDL number MFCD06364464
  • Cas Number : 793716-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11446
Availability details for
Quantity
  • Availability
  • Add to Cart