ENA048303483 Enamine

4-(2,4-difluorobenzenesulfonamido)butanoic acid

  • MDL number MFCD06366659
  • Cas Number : 794553-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11454
Availability details for
Quantity
  • Availability
  • Add to Cart