ENA048303484 Enamine

5-(diethylsulfamoyl)-2-(morpholin-4-yl)benzoic acid

  • MDL number MFCD06366735
  • Cas Number : 794554-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11455
Availability details for
Quantity
  • Availability
  • Add to Cart