ENA048303499 Enamine

2-chloro-N-[1-phenyl-3-(5,6,7,8-tetrahydronaphthalen-2-yl)-1H-pyrazol-5-yl]acetamide

  • MDL number MFCD06366762
  • Cas Number : 956778-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11490
Availability details for
Quantity
  • Availability
  • Add to Cart