ENA048303502 Enamine

2-[(4-nitrophenyl)thio]propanoic acid

  • MDL number MFCD06368074
  • Cas Number : 794582-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11493

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11493
Availability details for
Quantity
  • Availability
  • Add to Cart